CAS RN 10431-98-8
CAS RN 10431-98-8
IUPAC Name:
2-ethyl-4,5-dihydro-1,3-oxazole
Synonyms:
polyethyloxazoline
Molecular Weight (g/mol):
99.13
Molecular Formula:
C5H9NO
InChi Key:
NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES:
CCC1=NCCO1
1
–
3
of
3
results
3
2-Ethyl-2-oxazoline, 99+%, Thermo Scientific Chemicals
CAS: 10431-98-8 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00005302 InChI Key: NYEZZYQZRQDLEH-UHFFFAOYSA-N Synonym: 2-ethyl-2-oxazoline,2-ethyloxazoline,2-ethyl-4,5-dihydrooxazole,oxazole, 2-ethyl-4,5-dihydro,2-oxazoline, 2-ethyl,unii-b8cd92t4p5,unii-jm7uh9q3yn,unii-hnx7574gtx,unii-s5dpg6k67i,ethyl oxazoline PubChem CID: 66412 IUPAC Name: 2-ethyl-4,5-dihydro-1,3-oxazole SMILES: CCC1=NCCO1