accessibility menu, dialog, popup

Test

CAS RN 104197-15-1

CAS RN 104197-15-1

IUPAC Name: 3,5-difluoro-4-methoxybenzonitrile
Synonyms: 3,5-difluoro-4-methoxybenzonitrile
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H5F2NO
InChi Key: FTPRNOSXVHXNSB-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results