accessibility menu, dialog, popup

Test

CAS RN 104076-39-3

H2COHOH2COHONOHNNSOOHOOH

CAS RN 104076-39-3

IUPAC Name: N-(3-{3-[(piperidin-1-yl)methyl]phenoxy}propyl)-1,3-benzothiazol-2-amine; bis(formic acid); bis(prop-2-enoic acid)
Synonyms: bis(acrylic acid); bis(formic acid); zolantidine
Molecular Weight (g/mol): 617.71
Molecular Formula: C30H39N3O9S
InChi Key: LUWINHZYJPMGDV-UHFFFAOYSA-N
SMILES: OC=O.OC=O.OC(=O)C=C.OC(=O)C=C.C(CNC1=NC2=CC=CC=C2S1)COC1=CC=CC(CN2CCCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results