accessibility menu, dialog, popup

Test

CAS RN 104-97-2

ClO

CAS RN 104-97-2

IUPAC Name: 3-cyclopentylpropanoyl chloride
Synonyms: 3-cyclopentylpropanoyl chloride
Molecular Weight (g/mol): 160.64
Molecular Formula: C8H13ClO
InChi Key: SZQVEGOXJYTLLB-UHFFFAOYSA-N
SMILES: ClC(=O)CCC1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results