man

Test

CAS RN 104-92-7

H3COBr

CAS RN 104-92-7

IUPAC Name: 1-bromo-4-methoxybenzene
Synonyms: bromoanisole
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: QJPJQTDYNZXKQF-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)