accessibility menu, dialog, popup

Test

CAS RN 104-87-0

CH3O

CAS RN 104-87-0

IUPAC Name: 4-methylbenzaldehyde
Synonyms: P-tolualdehyde paratolualdehyde para-toluyl aldehyde PTAL
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: FXLOVSHXALFLKQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C=C1
Boiling Point 
  • (4)
  • (7)
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (3)
  • (4)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (1)
  • (1)
  • (4)
  • (7)
  • (7)
  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (7)