man

Test

CAS RN 104-78-9

H3CNCH3NH2

CAS RN 104-78-9

IUPAC Name: (3-aminopropyl)diethylamine
Synonyms: (3-aminopropyl)diethylamine
Molecular Weight (g/mol): 130.24
Molecular Formula: C7H18N2
InChi Key: QOHMWDJIBGVPIF-UHFFFAOYSA-N
SMILES: CCN(CC)CCCN
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)