accessibility menu, dialog, popup

Test

CAS RN 104-50-7

CH3OO

CAS RN 104-50-7

IUPAC Name: 5-butyloxolan-2-one
Synonyms: gamma-octalactone
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: IPBFYZQJXZJBFQ-UHFFFAOYNA-N
SMILES: CCCCC1CCC(=O)O1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)