accessibility menu, dialog, popup

Test

CAS RN 104-12-1

ClNCO

CAS RN 104-12-1

IUPAC Name: 1-chloro-4-isocyanatobenzene
Synonyms: 1-chloro-4-isocyanatobenzene
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: ADAKRBAJFHTIEW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)