man

Test

CAS RN 104-03-0

OHONOO

CAS RN 104-03-0

IUPAC Name: 2-(4-nitrophenyl)acetic acid
Synonyms: 2-(4-nitrophenyl)acetic acid P-nitrophenylacetic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: YBADLXQNJCMBKR-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)