man

Test

CAS RN 103986-53-4

CH3BHOOH

CAS RN 103986-53-4

IUPAC Name: (4-methylnaphthalen-1-yl)boronic acid
Synonyms: 4-methylnaphthalen-1-ylboronic acid
Molecular Weight (g/mol): 186.02
Molecular Formula: C11H11BO2
InChi Key: JHVQEUGNYSVSDH-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(C=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)