accessibility menu, dialog, popup

Test

CAS RN 10386-28-4

NNI

CAS RN 10386-28-4

IUPAC Name: 3-iodopyridine-4-carbonitrile
Synonyms: 3-iodopyridine-4-carbonitrile
Molecular Weight (g/mol): 230.01
Molecular Formula: C6H3IN2
InChi Key: KMFIQUYSWTUCGD-UHFFFAOYSA-N
SMILES: IC1=C(C=CN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results