CAS RN 10351-75-4
CAS RN 10351-75-4
IUPAC Name:
1H-1,3-benzodiazole-5,6-dicarboxylic acid
Synonyms:
1H-1,3-benzodiazole-5,6-dicarboxylic acid
Molecular Weight (g/mol):
206.16
Molecular Formula:
C9H6N2O4
InChi Key:
PIPQOFRJDBZPFR-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(C=C2N=CNC2=C1)C(O)=O
1
–
1
of
1
results
1
Benzimidazole-5,6-dicarboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 10351-75-4 Molecular Formula: C9H6N2O4 Molecular Weight (g/mol): 206.16 MDL Number: MFCD03093058 InChI Key: PIPQOFRJDBZPFR-UHFFFAOYSA-N Synonym: benzimidazole-5,6-dicarboxylic acid,1h-benzo d imidazole-5,6-dicarboxylic acid,1h-1,3-benzodiazole-5,6-dicarboxylic acid,1h-benzoimidazole-5,6-dicarboxylic,1h-benzoimidazole-5,6-dicarboxylic acid,acmc-1bsyc,5,6-benzimidazoledicarboxylic acid,benzo d imidazole-5,6-dicarboxylic acid PubChem CID: 7023591 IUPAC Name: 1H-benzimidazole-5,6-dicarboxylic acid SMILES: OC(=O)C1=C(C=C2N=CNC2=C1)C(O)=O