accessibility menu, dialog, popup

Test

CAS RN 103476-89-7

CH3H3CNHH3COOHNCH3CH3ONHHNNH2NH2O

CAS RN 103476-89-7

IUPAC Name: [amino({[(4S)-4-[(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methylpentanamido]-5-oxopentyl]amino})methylidene]azanium
Synonyms: [amino({[(4S)-4-[(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methylpentanamido]-5-oxopentyl]amino})methylidene]azanium
Molecular Weight (g/mol): 427.57
Molecular Formula: C20H39N6O4
InChi Key: GDBQQVLCIARPGH-ULQDDVLXSA-O
SMILES: CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=[NH2+])C=O
Form 
  • (5)
Product Type 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (5)