CAS RN 10343-06-3
CAS RN 10343-06-3
IUPAC Name:
[3,4,5-tris(acetyloxy)-6-hydroxyoxan-2-yl]methyl acetate
Synonyms:
[3,4,5-tris(acetyloxy)-6-hydroxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol):
348.3
Molecular Formula:
C14H20O10
InChi Key:
IEOLRPPTIGNUNP-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2,3,4,6-Tetra-O-acetyl-D-glucopyranose, 98%
CAS: 10343-06-3 Molecular Formula: C14H20O10 Molecular Weight (g/mol): 348.304 MDL Number: MFCD00061634 InChI Key: IEOLRPPTIGNUNP-RQICVUQASA-N Synonym: 2,3,4,6-tetra-o-acetyl-d-glucopyranose,2,3,4,6-tetra-o-acetylglucopyranose,2r,3r,4s,5r-3,4,5-tris acetyloxy-6-hydroxyoxan-2-yl methyl acetate,2,3,4,6-tetra-o-acetyl-beta-d-glucopyranose,pubchem10600,d-glucopyranose 2,3,4,6-tetraacetate,2,3,4,6-tetra-o-acetyl-glucopyranose,2,3,4,6-tetraacetate d-glucopyranose,d-glucopyranose, 2,3,4,6-tetraacetate,2r,3r,4s,5r-2-acetoxymethyl-6-hydroxytetrahydro-2h-pyran-3,4,5-triyl triacetate PubChem CID: 11824227 IUPAC Name: [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate SMILES: CC(=O)OCC1C(C(C(C(O1)O)OC(=O)C)OC(=O)C)OC(=O)C