accessibility menu, dialog, popup

Test

CAS RN 10342-59-3

CH3NOO

CAS RN 10342-59-3

IUPAC Name: 1-nitro-4-propylbenzene
Synonyms: 4-propylnitrobenzene
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: SXQBFCVVZIYXHV-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results