man

Test

CAS RN 10340-91-7

CN

CAS RN 10340-91-7

IUPAC Name: (isocyanomethyl)benzene
Synonyms: (isocyanomethyl)benzene
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: RIWNFZUWWRVGEU-UHFFFAOYSA-N
SMILES: [C-]#[N+]CC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)