CAS RN 1034-01-1
CAS RN 1034-01-1
IUPAC Name:
octyl 3,4,5-trihydroxybenzoate
Synonyms:
octyl gallate caprylyl gallate
Molecular Weight (g/mol):
282.34
Molecular Formula:
C15H22O5
InChi Key:
NRPKURNSADTHLJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
1
–
1
of
1
results
1
n-Octyl gallate, 98+%, Thermo Scientific Chemicals
CAS: 1034-01-1 Molecular Formula: C15H22O5 Molecular Weight (g/mol): 282.336 MDL Number: MFCD00002197 InChI Key: NRPKURNSADTHLJ-UHFFFAOYSA-N Synonym: octyl gallate,n-octyl gallate,progallin o,stabilizer ga 8,n-octylgallate,gallic acid, octyl ester,gallic acid n-octyl ester,gallic acid octyl ester,benzoic acid, 3,4,5-trihydroxy-, octyl ester,oktylester kyseliny gallove PubChem CID: 61253 ChEBI: CHEBI:83631 IUPAC Name: octyl 3,4,5-trihydroxybenzoate SMILES: CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O