accessibility menu, dialog, popup

Test

CAS RN 103249-79-2

CH3NNOHO

CAS RN 103249-79-2

IUPAC Name: 4-methyl-2-phenylpyrimidine-5-carboxylic acid
Synonyms: 4-methyl-2-phenylpyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: ZPXPNKCBUXSJFK-UHFFFAOYSA-N
SMILES: CC1=NC(=NC=C1C(O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results