accessibility menu, dialog, popup

Test

CAS RN 103213-32-7

OOSHNOO

CAS RN 103213-32-7

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoate
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoate
Molecular Weight (g/mol): 584.71
Molecular Formula: C37H30NO4S
InChi Key: KLBPUVPNPAJWHZ-UMSFTDKQSA-M
SMILES: [O-]C(=O)[C@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results