accessibility menu, dialog, popup

Test

CAS RN 103203-84-5

OOHO

CAS RN 103203-84-5

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-6-carboxylic acid
Synonyms: 3,4-dihydro-2H-1-benzopyran-6-carboxylic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: IFKANGOXGBPILW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results