man

Test

CAS RN 103-84-4

CH3ONH

CAS RN 103-84-4

IUPAC Name: N-phenylacetamide
Synonyms: acetanilide acetic acid anilide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: FZERHIULMFGESH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (5)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (5)
  • (5)
  • (2)
  • (1)