man

Test

CAS RN 103-78-6

CH3O

CAS RN 103-78-6

IUPAC Name: 1-cyclohexylpropan-2-one
Synonyms: cyclohexylacetone
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: UHBZEAPZATVYKV-UHFFFAOYSA-N
SMILES: CC(=O)CC1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)