accessibility menu, dialog, popup

Test

CAS RN 103-63-9

Br

CAS RN 103-63-9

IUPAC Name: (2-bromoethyl)benzene
Synonyms: (2-bromoethyl)benzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: WMPPDTMATNBGJN-UHFFFAOYSA-N
SMILES: BrCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)