accessibility menu, dialog, popup

Test

CAS RN 103-28-6

H3CCH3OO

CAS RN 103-28-6

IUPAC Name: benzyl 2-methylpropanoate
Synonyms: benzyl isobutyrate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: UIKJRDSCEYGECG-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results