man

Test

CAS RN 10294-39-0

H2OH2OH2OOClOOOOClOOOBa

CAS RN 10294-39-0

IUPAC Name: barium(2+) trihydrate diperchlorate
Synonyms: barium(2+) trihydrate diperchlorate
Molecular Weight (g/mol): 390.26
Molecular Formula: BaCl2H6O11
InChi Key: NBHDMFAHVJWOHD-UHFFFAOYSA-L
SMILES: O.O.O.[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)