accessibility menu, dialog, popup

Test

CAS RN 1027511-41-6

H3COONNNBr

CAS RN 1027511-41-6

IUPAC Name: ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
Synonyms: ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
Molecular Weight (g/mol): 270.09
Molecular Formula: C9H8BrN3O2
InChi Key: PZZTXIWJXOMKEU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2N=CC(Br)=CN2N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results