accessibility menu, dialog, popup

Test

CAS RN 10250-64-3

CH3NNOOH

CAS RN 10250-64-3

IUPAC Name: 1-methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
Synonyms: 2-methyl-5-phenylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: RTBQZMPYPFQBHA-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results