accessibility menu, dialog, popup

Test

CAS RN 10212-25-6

HOOONNHNHOHCl

CAS RN 10212-25-6

IUPAC Name: hydrogen (2R,4R,5R,6S)-4-(hydroxymethyl)-10-imino-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-5-ol chloride
Synonyms: hydrogen (2R,4R,5R,6S)-4-(hydroxymethyl)-10-imino-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-5-ol chloride
Molecular Weight (g/mol): 261.66
Molecular Formula: C9H12ClN3O4
InChi Key: KZOWNALBTMILAP-JBMRGDGGSA-N
SMILES: [H+].[Cl-].OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results