man

Test

CAS RN 102-36-3

ClClNCO

CAS RN 102-36-3

IUPAC Name: 1,2-dichloro-4-isocyanatobenzene
Synonyms: 3,4-dichlorphenylisocyanate
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: MFUVCHZWGSJKEQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results