man

Test

CAS RN 102-04-5

O

CAS RN 102-04-5

IUPAC Name: 1,3-diphenylpropan-2-one
Synonyms: dibenzyl ketone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: YFKBXYGUSOXJGS-UHFFFAOYSA-N
SMILES: O=C(CC1=CC=CC=C1)CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)