accessibility menu, dialog, popup

Test

CAS RN 101990-72-1

NClClCl

CAS RN 101990-72-1

IUPAC Name: 2,6-dichloro-4-(chloromethyl)pyridine
Synonyms: 2,6-dichloro-4-(chloromethyl)pyridine
Molecular Weight (g/mol): 196.46
Molecular Formula: C6H4Cl3N
InChi Key: UYBVOPLURVHJOX-UHFFFAOYSA-N
SMILES: ClCC1=CC(Cl)=NC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results