man

Test

CAS RN 1019008-21-9

CH3H3CCH3OONN

CAS RN 1019008-21-9

IUPAC Name: tert-butyl 3-(pyrrolidin-1-yl)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(pyrrolidin-1-yl)azetidine-1-carboxylate
Molecular Weight (g/mol): 226.32
Molecular Formula: C12H22N2O2
InChi Key: YQSWCTNNHKPDNK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(C1)N1CCCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results