accessibility menu, dialog, popup

Test

CAS RN 101712-20-3

CH3OFF

CAS RN 101712-20-3

IUPAC Name: 1-(2,6-difluorophenyl)propan-2-one
Synonyms: 1-(2,6-difluorophenyl)propan-2-one
Molecular Weight (g/mol): 170.16
Molecular Formula: C9H8F2O
InChi Key: LAWHOFKPDMZDLJ-UHFFFAOYSA-N
SMILES: CC(=O)CC1=C(F)C=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results