CAS RN 10170-68-0
CAS RN 10170-68-0
IUPAC Name:
tris(oxolane); trichlorochromium
Synonyms:
tris(THF); chromium chloride
Molecular Weight (g/mol):
374.67
Molecular Formula:
C12H24Cl3CrO3
InChi Key:
CYOMBOLDXZUMBU-UHFFFAOYSA-K
SMILES:
Cl[Cr](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
1
–
1
of
1
results
1
Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific Chemicals
CAS: 10170-68-0 Molecular Formula: C12H24Cl3CrO3 Molecular Weight (g/mol): 374.67 MDL Number: MFCD00013222 InChI Key: CYOMBOLDXZUMBU-UHFFFAOYSA-K Synonym: chromium iii chloride tetrahydrofuran complex 1:3,chromium iii chloride tetrahydrofuran adduct,crcl3 thf 3,crcl3 3thf,chromium iii chloride tetrahydrofuran complex,chromium chloride; tris tetrahydrofuran,chromium iii chloride complex 1:3 tetrahydrofuran PubChem CID: 11100829 IUPAC Name: oxolane;trichlorochromium SMILES: C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl