CAS RN 101691-94-5
CAS RN 101691-94-5
IUPAC Name:
4-(iodomethyl)oxane
Synonyms:
4-(iodomethyl)oxane
Molecular Weight (g/mol):
226.06
Molecular Formula:
C6H11IO
InChi Key:
MQLFSPBSNWUXSO-UHFFFAOYSA-N
SMILES:
ICC1CCOCC1
1
–
2
of
2
results
1
4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Thermo Scientific™
CAS: 101691-94-5 Molecular Formula: C6H11IO Molecular Weight (g/mol): 226.057 InChI Key: MQLFSPBSNWUXSO-UHFFFAOYSA-N Synonym: 4-iodomethyl tetrahydro-2h-pyran,4-iodomethyl oxane,4-iodomethyl-tetrahydro-pyran,4-iodomethyl tetrahydropyran,2h-pyran, tetrahydro-4-iodomethyl,4-iodomethyl-tetrahydro-2h-pyran,2h-pyran,tetrahydro-4-iodomethyl,acmc-20a1ce,4-iodomethyltetrahydropyran,4-iodomethyl-tetrahydropyran PubChem CID: 2795507 IUPAC Name: 4-(iodomethyl)oxane SMILES: C1COCCC1CI