man

Test

CAS RN 10139-47-6

ZnII

CAS RN 10139-47-6

IUPAC Name: zinc(2+) diiodide
Synonyms: zinc iodide
Molecular Weight (g/mol): 319.19
Molecular Formula: I2Zn
InChi Key: UAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES: [Zn++].[I-].[I-]
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)