CAS RN 1013-25-8
CAS RN 1013-25-8
IUPAC Name:
4-(2,5-dimethylphenyl)piperazin-1-ium
Synonyms:
4-(2,5-dimethylphenyl)piperazin-1-ium
Molecular Weight (g/mol):
191.3
Molecular Formula:
C12H19N2
InChi Key:
YRIFWVMRUFKWLM-UHFFFAOYSA-O
SMILES:
CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1
1
–
1
of
1
results
1
1-(2,5-Dimethylphenyl)piperazine, 96%, Thermo Scientific™
CAS: 1013-25-8 Molecular Formula: C12H19N2 Molecular Weight (g/mol): 191.30 MDL Number: MFCD00038378 InChI Key: YRIFWVMRUFKWLM-UHFFFAOYSA-O Synonym: 1-2,5-dimethylphenyl piperazine,1-2,5-xylyl piperazine,piperazine, 1-2,5-dimethylphenyl,2,5-dimethylphenyl piperazine,acmc-1bpa9,2,5-dimethylphenylpiperazine,n-2,5-dimethylphenyl piperazine,1-2,5-dimethylphenyl-piperazine,4-2,5-dimethyl-phenyl piperazine,1-2,5-dimethyl-phenyl-piperazine PubChem CID: 70542 IUPAC Name: 1-(2,5-dimethylphenyl)piperazine SMILES: CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1