accessibility menu, dialog, popup

Test

CAS RN 10102-94-0

H3CNBrO

CAS RN 10102-94-0

IUPAC Name: 5-bromo-1-methyl-1H-indole-3-carbaldehyde
Synonyms: 5-bromo-1-methylindole-3-carbaldehyde
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: NZJJLQUTQOICBN-UHFFFAOYSA-N
SMILES: CN1C=C(C=O)C2=CC(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results