accessibility menu, dialog, popup

Test

CAS RN 10102-68-8

CAS RN 10102-68-8

IUPAC Name: calcium diiodide
Synonyms: calcium iodide
Molecular Weight (g/mol): 293.89
Molecular Formula: CaI2
InChi Key: UNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES: [Ca++].[I-].[I-]
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)