accessibility menu, dialog, popup

Test

CAS RN 10102-20-2

CAS RN 10102-20-2

IUPAC Name: disodium pentahydrate tellurate
Synonyms: sodium tellurite
Molecular Weight (g/mol): 327.65
Molecular Formula: H10Na2O9Te
InChi Key: OVBVNEXDNNHXIU-UHFFFAOYSA-L
SMILES: O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results