accessibility menu, dialog, popup

Test

CAS RN 10102-18-8

CAS RN 10102-18-8

IUPAC Name: disodium selenite
Synonyms: sodium selenite disodium selenite natriumselenit
Molecular Weight (g/mol): 172.95
Molecular Formula: Na2O3Se
InChi Key: BVTBRVFYZUCAKH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][Se]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (5)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)