man

Test

CAS RN 101-83-7

HN

CAS RN 101-83-7

IUPAC Name: N-cyclohexylcyclohexanamine
Synonyms: dicha N,N-diclohexylamine dodecahydrodiphenylamine dicyklohexylamin
Molecular Weight (g/mol): 181.32
Molecular Formula: C12H23N
InChi Key: XBPCUCUWBYBCDP-UHFFFAOYSA-N
SMILES: C1CCC(CC1)NC1CCCCC1
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)