accessibility menu, dialog, popup

Test

CAS RN 100905-33-7

H3COONOOOCH3OCH3

CAS RN 100905-33-7

IUPAC Name: ethyl 4,5-dimethoxy-2-nitrobenzoate
Synonyms: ethyl 4,5-dimethoxy-2-nitrobenzoate
Molecular Weight (g/mol): 255.23
Molecular Formula: C11H13NO6
InChi Key: VNPCXITWHOOQAJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(OC)=C(OC)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results