CAS RN 100846-24-0
CAS RN 100846-24-0
IUPAC Name:
5-(trifluoromethyl)-1H-indole
Synonyms:
5-(trifluoromethyl)-1H-indole
Molecular Weight (g/mol):
185.15
Molecular Formula:
C9H6F3N
InChi Key:
LCFDJWUYKUPBJM-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C2NC=CC2=C1
1
–
1
of
1
results
1
5-(Trifluoromethyl)indole, 98%, Thermo Scientific Chemicals
CAS: 100846-24-0 Molecular Formula: C9H6F3N Molecular Weight (g/mol): 185.149 MDL Number: MFCD03095341 InChI Key: LCFDJWUYKUPBJM-UHFFFAOYSA-N Synonym: 5-trifluoromethyl indole,5-trifluoromethyl-1h-indole,5-trifluoromethylindole,1h-indole, 5-trifluoromethyl,zlchem 948,pubchem1712,acmc-1box4,5-trifluoromethyl-indole PubChem CID: 2763075 IUPAC Name: 5-(trifluoromethyl)-1H-indole SMILES: C1=CC2=C(C=CN2)C=C1C(F)(F)F