accessibility menu, dialog, popup

Test

CAS RN 10070-92-5

CAS RN 10070-92-5

IUPAC Name: pyrimidine-5-carbaldehyde
Synonyms: pyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 108.1
Molecular Formula: C5H4N2O
InChi Key: FREJAOSUHFGDBW-UHFFFAOYSA-N
SMILES: O=CC1=CN=CN=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)