CAS RN 1007-28-9
CAS RN 1007-28-9
IUPAC Name:
6-chloro-N2-ethyl-1,3,5-triazine-2,4-diamine
Synonyms:
deisopropylatrazine 6-deisopropylatrazine caswell no. 033F C5H8ClN5 desisopropylatrazine
Molecular Weight (g/mol):
173.6
Molecular Formula:
C5H8ClN5
InChi Key:
IVENSCMCQBJAKW-UHFFFAOYSA-N
SMILES:
CCNC1=NC(N)=NC(Cl)=N1
1
–
5
of
5
results