accessibility menu, dialog, popup

Test

CAS RN 100643-71-8

ClNHN

CAS RN 100643-71-8

IUPAC Name: 13-chloro-2-(piperidin-4-ylidene)-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Synonyms: desloratadine desloratidine neoclarityn descarboethoxyoratidine aerius denosin clarinex claramax descarboethoxyloratadine clarinex reditabs
Molecular Weight (g/mol): 310.83
Molecular Formula: C19H19ClN2
InChi Key: JAUOIFJMECXRGI-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(CCC3=CC=CN=C3C2=C2CCNCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results