accessibility menu, dialog, popup

Test

CAS RN 100641-16-5

NOOONOOONOOOLu

CAS RN 100641-16-5

IUPAC Name: lutetium(3+) hydrate trinitrate
Synonyms: lutetium(3+) hydrate trinitrate
Molecular Weight (g/mol): 378.99
Molecular Formula: H2LuN3O10
InChi Key: XZBUOUZGWUJJES-UHFFFAOYSA-N
SMILES: O.[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)