accessibility menu, dialog, popup

Test

CAS RN 1006-94-6

CAS RN 1006-94-6

IUPAC Name: 5-methoxy-1H-indole
Synonyms: 5-methoxyindole
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: DWAQDRSOVMLGRQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC=CC2=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)